Learn More
Invitrogen™ PureLink™ PCR Micro Kit
PureLink PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions.
62.75 € - 600.00 €
Specifications
Content And Storage | Kit includes columns, binding and wash buffers, recovery tubes, and the manual. Store at room temperature. |
---|---|
Format | Spin Column |
Elution Volume Max | 20 μL |
Elution Volume Min | 5 μL |
High-throughput Compatibility | Not High-throughput Compatible (Manual) |
Includes: Columns, binding and wash buffers, recovery tubes, and the manual
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10409033
|
Invitrogen™
K310050 |
50 Preps |
140.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10490344
|
Invitrogen™
K310010 |
10 Preps |
62.75 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10043063
|
Invitrogen™
K310250 |
250 Preps |
600.00 €
250 preps |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PureLink™ PCR Micro Kits are ideal for purifying and concentrating DNA from low-yielding PCR or restriction digestion reactions.These highly purified DNA fragments and amplicons provide excellent results in downstream applications such as DNA sequencing, restriction digestion, cloning, PCR, and labeling. PureLink™ PCR Micro Kits provide a simple and rapid method for purifying and concentrating DNA fragments ranging in size from 100 bp-12 kb.
The unique micro-nozzle design allows up to 95% recovery of DNA fragments with as little as a 5μL elution volume. With less than a 10% coefficient of variance in the elution volume you can be sure that no ethanol is carried over from the wash steps giving you the best chance of success in downstream applications. Removal of primers, dNTPs, enzymes, and salts exceeds 98.5%. This kit can also efficiently remove unincorporated dye-labeled nucleotides from a cDNA labeling reaction.
Not High Throughput-Compatible (Manual)
Order Info
Shipping Condition: Room Temperature
Specifications
Kit includes columns, binding and wash buffers, recovery tubes, and the manual. Store at room temperature. | |
20 μL | |
Not High-throughput Compatible (Manual) | |
PCR Micro Kit | |
100 bp to 12 kb | |
125 μL | |
Cloning, Standard PCR |
Spin Column | |
5 μL | |
PureLink™ | |
10 min. | |
Room Temperature | |
DNA from Enzymatic Reactions (e.g. PCR) | |
Silica Spin Column |
Safety and Handling
- PCR Micro Kit
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Guanidine hydrochloride
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.