Learn More
Invitrogen™ SUPERase·In™ RNase Inhibitor (20 U/μL)
Binds and inactivates a wide variety of RNases
Brand: Invitrogen™ AM2696
554.80 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Includes: Supplied in a single tube
Description
SUPERase·In™ RNase Inhibitor is a protein-based inhibitor of nonhuman origin that noncovalently binds and inhibits the most common and troublesome RNases, including RNase A, B, C, 1, and T1. SUPERase·In RNase Inhibitor can be used in any application where RNase contamination could be problematic. Because it inhibits a broader range of RNases than traditional RNase inhibitors, SUPERase·In is the most effective RNase inhibitor available, providing a higher level of protection against degradation. SUPERase·In does not interfere with other enzymes such as RNA polymerases, reverse transcriptase, or Taq DNA polymerase. Additionally, SUPERase In is active up to 65°C and over a pH range of 5.5 to 8.5.
- Completely removes RNase contamination from glass and plastic surfaces
- Excels at removing high levels of RNase contamination where similar products fail
- Proven effective at removing high concentrations of dried-on RNase A
- Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free
SUPERase·In RNase Inhibitor at 1U/μL will block the degradation of 0.1μg/μL labeled RNA by 2.5pg/μL of RNase A, 2.5pg/μL of RNase I, and 0.0075U/μL of RNase T1, for four hours at 37°C, in 20mM Tris-HCl, pH 7.5, 50mM NaCl, 1mM EDTA. Analysis is by denaturing PAGE. SUPERase·In is currently the only ribonuclease inhibitor for which the unit activity is defined by such a functional assay.
If using this product with standard antibody purification methods, please contact technical support to discuss experimental design.
Gene Expression Analysis and Genotyping, In Vitro Transcription, PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription
Order Info
Shipping Condition: Dry ice

Specifications
50 mM | |
Ambion™, SUPERase•In™ | |
Dry Ice |
Supplied in a single tube that shouldbe stored at -20°C. Storage buffer: 2 mM KH2PO4, 8 mM Na2HPO4,2.7 mM KCl, 137 mM NaCl, pH 7.4 in 50% glycerol |
|
10,000 U |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.