missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sf9 Cells in Grace's

Derived from the USDA parental IPLBSF-21 (Sf21) cell line, derived from the pupal ovarian tissue of the fall army worm, Spodoptera frugiperda

Brand:  Invitrogen™ B82501

588.60 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10503433

  • 654.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

Review the Material Safety Data Sheet before handling.

This item is not returnable. View return policy

Description

Description

Gibco Sf9 cells are commonly used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins.

  • Recombinant protein expression from a variety of expression systems
  • Good growth in adherent or suspension culture
  • Small, regular size that generates even monolayers and plaques
  • Quality and performance testing

Baculovirus Production and Titering, Bioproduction, Cell Culture, Insect Cell Culture, Insect Expression, Insect Expression Cell Lines, Protein Expression, Proteins, Expression, Isolation and Analysis, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: Liquid nitrogen (vapor phase)
Shipping conditions: Frozen
Insect Cells
Insect Cells
Sf9
S. frugiperda
1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Sf9 Cells in Grace's > 1mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for any animal or human therapeutic or diagnostic use.