missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ III Reverse Transcriptase

Invitrogen SuperScript III Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) created by the introduction of several mutations for reduced RNase H activity, increased half-life, and improved thermal stability.

Brand:  Invitrogen™ 18080044

 View more versions of this product

Product Code. 10368252

  • 450.00 € / 10ku

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen SuperScript III Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) created by the introduction of several mutations for reduced RNase H activity, increased half-life, and improved thermal stability. SuperScript III RT offers higher cDNA yields, improved cDNA lengths, improved efficiency on GC-rich target RNAs, and overall better performance than wild-type MMLV and MMLV RNase H-minus enzymes.

SuperScript RTs are the most highly trusted and widely used RTs with over 50,000 citations, reviews, and publications to date. The enzyme is purified to near homogeneity from E. coli containing the modified pol gene of Moloney Murine Leukemia Virus.

Features of SuperScript III Reverse Transcriptase include:
• First-strand cDNA may be synthesized at temperatures up to 55°C
• cDNA can be generated from 100 bp to >12 kb
• Increased specificity
• High yields of cDNA and full-length products

Note: The latest member of the SuperScript RT family, SuperScript IV Reverse Transcriptase (Cat. No. 18090010), features enhanced thermostability, processivity, yields, and performance with any RNA samples, including those of suboptimal purity or integrity.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• SuperScript™ III Reverse Transcriptase, 50 μL (10,000 units at 200 U/μL)
• 5X First-strand buffer, 1 mL
• DTT, 500 μL (100 mM)

Store at –20°C.

High
Reverse Transcription
10,000 Units
Reduced
200 U/μL
50 Reactions
Reverse Transcription
RNA
Stand-alone Enzyme
30 to 50 min.
50°C
SuperScript III
Dry Ice
First-Strand cDNA
Separate components
Up to 12.3 kb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ SuperScript™ III Reverse Transcriptase > 50 Reactions; 10,000 units

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.