missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SuperScript™ III Reverse Transcriptase

Invitrogen SuperScript III Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) created by the introduction of several mutations for reduced RNase H activity, increased half-life, and improved thermal stability.

144.00 € - 1562.00 €

Specifications

Format Stand-alone Enzyme
GC-Rich PCR Performance High
Reaction Speed 30 to 50 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C
View More Specs

Products 3
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
10432122
View Documents
Invitrogen™
18080093
10 Reactions
144.00 €
2ku
Please to purchase this item. Need a web account? Register with us today!
10368252
View Documents
Invitrogen™
18080044
50 Reactions
441.00 €
10ku
Please to purchase this item. Need a web account? Register with us today!
12087539
View Documents
Invitrogen™
18080085
200 Reactions
1562.00 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Invitrogen SuperScript III Reverse Transcriptase is a genetically engineered MMLV reverse transcriptase (RT) created by the introduction of several mutations for reduced RNase H activity, increased half-life, and improved thermal stability. SuperScript III RT offers higher cDNA yields, improved cDNA lengths, improved efficiency on GC-rich target RNAs, and overall better performance than wild-type MMLV and MMLV RNase H-minus enzymes.

SuperScript RTs are the most highly trusted and widely used RTs with over 50,000 citations, reviews, and publications to date. The enzyme is purified to near homogeneity from E. coli containing the modified pol gene of Moloney Murine Leukemia Virus.

Features of SuperScript III Reverse Transcriptase include:
• First-strand cDNA may be synthesized at temperatures up to 55°C
• cDNA can be generated from 100 bp to >12 kb
• Increased specificity
• High yields of cDNA and full-length products

Note: The latest member of the SuperScript RT family, SuperScript IV Reverse Transcriptase (Cat. No. 18090010), features enhanced thermostability, processivity, yields, and performance with any RNA samples, including those of suboptimal purity or integrity.

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

Stand-alone Enzyme
30 to 50 min.
50°C
Reduced
200 U/μL
Separate components
Up to 12.3 kb
High
Reverse Transcription
SuperScript III
Dry Ice
First-Strand cDNA
Reverse Transcription
RNA
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.