missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Total Exosome Isolation Reagent (from serum)

Total Exosome Isolation Reagent (from serum) enables fast and efficient enrichment of intact exosomes from blood serum samples with our simple and reliable protocol.

Brand:  Invitrogen™ 4478360

Product Code. 13355394

  • 397.00 € / 6mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Total Exosome Isolation Reagent (from serum) enables fast and efficient enrichment of intact exosomes from blood serum samples with our simple and reliable protocol.

  • Maximise recovery of intact exosomes for any type of downstream applications
  • Maximize flexibility—can be scaled up or down depending on the sample size
  • Enables simple and reliable concentration of intact exosomes from blood serum samples

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2°C to 8°C.
Exosomes
Serum
Total Exosome Isolation
6 mL
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Total Exosome Isolation Reagent (from serum) > 6mL; from serum

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.