missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Total Exosome RNA & Protein Isolation Kit

The Total Exosome RNA and Protein Isolation Kit enables isolation of total RNA and proteins from exosome samples extracted from biological fluids (e.g., by using a Total Exosome Isolation (from cell culture media) or Total Exosome Isolation (from serum).

Brand:  Invitrogen™ 4478545

413.95 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.



Product Code. 13463119

  • 487.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Total Exosome RNA and Protein Isolation Kit enables isolation of total RNA and proteins from exosome samples extracted from biological fluids (e.g., by using a Total Exosome Isolation (from cell culture media) or Total Exosome Isolation (from serum).

  • Extracts highly pure total RNA (including small RNA fraction) from exosomes
  • Recovers protein and RNA from the same sample
  • Compatible with all protocols for exosome isolation

Not high-throughput compatible.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 2°C to 8°C.
Total Exosome Isolation
Not High-throughput Compatible (Manual)
Total RNA, Protein, RNA & Protein, miRNA
Organelle & Vesicle-Related Samples, Exosomes
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR), Western Blot, microRNA Analysis
Total RNA, Protein, micro RNA
Spin Column (Glass Fiber Filter), Organic Extraction
40 preps
Room Temperature
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Total Exosome RNA & Protein Isolation Kit

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : .alpha.-[4-(1,1,3,3-tetramethylbutyl)phenyl]-.omega.-hydroxy-poly(oxy-1,2-ethanediyl)

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.