missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ SSC, 20X
GREENER_CHOICE

For use in nucleic acid hybridizations and blot transfer applications

32.41 € - 93.00 €

Specifications

Content And Storage Store at room temperature.
Concentration 20 X
Product Type SSC
Form Liquid
For Use With (Application) Nucleic Acid Gel Electrophoresis, Blotting
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10515203
GREENER_CHOICE
View Documents
Invitrogen™
15557044
1 L
32.41 €
1 L
In Stock
to see stock.
Edge
Add to basket
 
11598976
View Documents
Invitrogen™
15557036
4 L
93.00 €
4 L
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

UltraPure 20X SSC is a solution formulated and used in concentrations ranging from 0.2X to 20X, depending on the application.

  • Supplied in 1L plastic bottles/4L/10L stackable Cubitainer™ Box
  • Sterile-filtered UltraPure 20X SSC contains 3.0 M NaCl and 0.3 M sodium citrate, at pH 7.0
  • Performance and Quality Testing: No DNase or RNase activity detected

DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, Microarray Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis & Blotting, Southern Blotting

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Store at room temperature.
SSC
Nucleic Acid Gel Electrophoresis, Blotting
Room Temperature
20 X
Liquid
UltraPure™
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.