Learn More
Invitrogen™ UltraPure™ TAE Buffer, 10X

Compatible with agarose gels
Brand: Invitrogen™ 15558042
25.02 EUR valid until 2025-12-31
Use promo code "24339" to get your promotional price.
Description
- UltraPure 10X TAE Buffer is sterile-filtered solution of 400 mM Tris-acetate and 10 mM EDTA
- TAE Buffer is most commonly used buffer for agarose DNA electrophoresis
- Supplied in 1 L plastic bottles/4 L/10 L stackable Cubitainer™ Box
- 1X TAE Buffer solution contains 40 mM Tris-acetate and 1mM EDTA at pH 8.3
- Performance and Quality Testing: No DNase activity detected. Sustainable packaging
Agarose Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Condition: Room Temperature

Specifications
Running Buffer | |
Store at room temperature. | |
Agarose Gels | |
UltraPure™ | |
1 L |
10 X | |
Bottle | |
Nucleic Acid Gel Electrophoresis, Blotting | |
TAE Buffer | |
Room Temperature |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2731534 | Certificate of Analysis | 29/01/2025 | 15558042 |
2651465 | Certificate of Analysis | 29/01/2025 | 15558042 |
2424468 | Certificate of Analysis | 29/01/2025 | 15558042 |
2424470 | Certificate of Analysis | 29/01/2025 | 15558042 |
2434098 | Certificate of Analysis | 29/01/2025 | 15558042 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.