missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents
Brand: Fisher BioReagents BP1335-1
This item is not returnable.
View return policy
Description
- Save time and simplify your buffer preparation step by using Fisher BioReagents 10X TAE buffer solution - simply dilute as needed
- 10X solution contains 0.4M tris-acetate and 0.01M EDTA
- To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TAE Solution with 9 volumes of ultrapure water, such as BP2819
- Tested for the absence of DNase, RNase, and protease
- Filtered, autoclaved, and functionally tested for use in electrophoresis applications
- TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
- Intended for laboratory research use only

Specifications
Tris-Acetate-EDTA | |
64-19-7, 7732-18-5, 60-00-4, 77-86-1 | |
1 L | |
Filtered through a 0.2-micron filter. | |
Nalgene Poly Bottle | |
TAE | |
Tris (<5.0%), Acetic Acid (1.0%), and EDTA (<1%). |
10X Solution | |
Liquid | |
DNase free | |
DNase-, RNase- and Protease-Free | |
Protease free | |
Electrophoresis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TAE Buffer, Tris-Acetate-EDTA, 10X Solution, Electrophoresis, Fisher BioReagents > Nalgene Poly Bottle; 1L
Spot an opportunity for improvement?Share a Content Correction