Learn More
Invitrogen™ UltraPure™ TBE Buffer, 10X
![GREENER_CHOICE](/content/dam/fishersci/glyphs/green_glyph.png)
Compatible with agarose gels
27.76 € - 194.00 €
Specifications
Buffer | Running Buffer |
---|---|
Concentration | 10 X |
Format | Bottle |
Gel Compatibility | Polyacrylamide Gels, Agarose Gels |
For Use With (Application) | Nucleic Acid Gel Electrophoresis, Blotting |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10358142
![]() |
Invitrogen™
15581044 |
1 L |
27.76 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11508986
|
Invitrogen™
15581028 |
10 L |
194.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UltraPure 10X TBE Buffer is a sterile-filtered solution of 1M Tris, 0.9 M boric acid, and 0.01M EDTA used to prepare 1X buffer for polyacrylamide and agarose gel electrophoresis.
- UltraPure 10X TBE Buffer is available in 1 L plastic bottle/10 L Cubitainer™ box
- Performance and quality testing: No DNase, RNase, or protease activity detected. Sustainable packaging
Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nucleic Acid Gel Electrophoresis & Blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Running Buffer | |
Bottle | |
Nucleic Acid Gel Electrophoresis, Blotting | |
TBE Buffer |
10 X | |
Polyacrylamide Gels, Agarose Gels | |
UltraPure™ | |
Room Temperature |
Safety and Handling
- TBE BUFFER, 10X
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1
Hazard Statement
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : boric acid
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.