missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast Nitrogen Base

Base medium for preparation of minimal and synthetic defined yeast media

Brand:  Invitrogen™ Q30007

161.10 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


Product Code. 10582893

  • 182.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Each pouch contains reagents to prepare 500mL of 10X YNB for Pichiamedia or 1000mL of 10X YNB for S. cerevisiaemedia.
This item is not returnable. View return policy

Description

Description

  • Contains ammonium sulfate but does not contain amino acids
  • Powder

Cloning, Transformation

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Each pouch contains 67 g of reagent to prepare 500 mL of 10X YNB for Pichia media or 1,000 mL of 10X YNB for S. cerevisiae media.

Store at room temperature. Guaranteed stable for 6 months when properly stored.
Microbiology Media
1 Pouch
Powder
P. pastoris, S. cerevisiae
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Yeast Nitrogen Base > 1 pouch

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.