Learn More
Iodine, 0.1 N standard solution, Thermo Scientific Chemicals
61.10 € - 95.60 €
Chemical Identifiers
CAS | 7553-56-2, 7681-11-0, 7732-18-5 |
---|---|
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
Synonym | iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
IUPAC Name | diiodine |
SMILES | II |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10123510
|
Thermo Scientific Acros
124220010 |
1 L | Glass bottle |
61.10 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10618693
|
Thermo Scientific Acros
124220025 |
2.5 L | Glass bottle |
95.60 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Volumetric standard solutionChemical Identifiers
7553-56-2, 7681-11-0, 7732-18-5 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
807 | |
diiodine |
I2 | |
MFCD00011355 MFCD00164163 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
CHEBI:17606 | |
II |
Specifications
7553-56-2, 7681-11-0, 7732-18-5 | |
Liquid | |
I2 | |
01,495; 02,200; 03,159; 04,258; 05,346; 06,293; 07,179; 08,256; 09,248; 10,210; 11,261; 12,235; 13,133; 14,181; 15,172; 16,182; 17,153 | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
0.0998 to 0.1002N (20°C) | |
Iodine, 0.1N Standard Solution |
Brown | |
I2 | |
MFCD00011355 MFCD00164163 | |
15, 5057 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
diiodine | |
807 | |
253.81 | |
1.0200g/mL |
Safety and Handling
GHS H Statement:
Harmful to aquatic life with long lasting effects.
GHS P Statement:
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 231-442-4
missing translation for 'rtecsNumber' : NN1575000
missing translation for 'tsca' : TSCA
- Iodine, 0.1N standard solution
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P314-Get medical advice/attention if you feel unwell.
RUO – Research Use Only