missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine solution, Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N), Honeywell Fluka™
Volumetric, Reag. Ph. Eur., 0.05 M I2 (0.1N)
130.38 € - 454.32 €
Chemical Identifiers
CAS | 7553-56-2, 7681-11-0, 7732-18-5 |
---|---|
Molecular Formula | I2 |
Molecular Weight (g/mol) | 253.81 |
MDL Number | MFCD00011355 MFCD00164163 |
InChI Key | PNDPGZBMCMUPRI-UHFFFAOYSA-N |
PubChem CID | 807 |
ChEBI | CHEBI:17606 |
SMILES | II |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15674790
|
Honeywell-Fluka
35090-2.5L |
2.5 L |
130.38 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15694790
|
Honeywell-Fluka
35090-5L |
5 L | |||||||
15604800
|
Honeywell-Fluka
35090-6X1L |
6 x 1 L |
293.36 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15684790
|
Honeywell-Fluka
35090-4X2.5L |
4 x 2.5 L |
454.32 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
7553-56-2, 7681-11-0, 7732-18-5 | |
253.81 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
CHEBI:17606 |
I2 | |
MFCD00011355 MFCD00164163 | |
807 | |
II |
Specifications
7553-56-2, 7681-11-0, 7732-18-5 | |
MFCD00011355 MFCD00164163 | |
3587194 | |
PNDPGZBMCMUPRI-UHFFFAOYSA-N | |
molecular iodine | |
807 | |
253.81g/mol | |
Iodine solution |
I2 | |
NONH for all modes of transport | |
iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode | |
II | |
253.81 | |
CHEBI:17606 | |
0.05 M |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Iodine solution
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : iodine