missing translation for 'onlineSavingsMsg'
Learn More
Iodine Solution, 0.05M, Fisher Chemical™ Product Code.: 12980754
compliance-icons compliance-icons

Iodine Solution, 0.05M, Fisher Chemical™
BIOPHARMA

Product Code. 12980754
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 12980754

Brand: Fisher Chemical SOL 114 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Quantity 100 mL
Concentration or Composition (by Analyte or Components) 0.05mol/L

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1097793 Certificate TypeCertificate of Analysis Date30/03/2025 Catalog NumberSOL 114 100
Lot Number1070448 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberSOL 114 100
Lot Number1001272 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberSOL 114 100
Lot Number1044832 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberSOL 114 100
Lot Number1036187 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberSOL 114 100
compliance-icons compliance-icons
Product Identifier
  • Iodine 0.5M Volumetric solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
Precautionary Statement
  • P264-Wash hands, forearms andface thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P314-Get medical advice/attention if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : iodine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iodine Solution, 0.05M, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.