missing translation for 'onlineSavingsMsg'
Learn More

Iodobenzene diacetate, 98%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 3240-34-4
Molecular Formula C10H11IO4
Molecular Weight (g/mol) 322.09
InChI Key ZBIKORITPGTTGI-UHFFFAOYSA-N
Synonym iodobenzene diacetate, diacetoxyiodo benzene, iodosobenzene diacetate, phenyliodine diacetate, phenyliodo diacetate, phenyliodoso acetate, phenyliodosodiacetate, phenyliodoso diacetate, bis acetato phenyliodine, benzene, diacetoxyiodo
PubChem CID 76724
IUPAC Name [acetyloxy(phenyl)-$l^{3}-iodanyl] acetate
SMILES CC(=O)OI(C1=CC=CC=C1)OC(=O)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10742411
View Documents
Thermo Scientific Acros
176560250
25 g Glass bottle N/A
N/A
10265750
View Documents
Thermo Scientific Acros
176561000
100 g Glass bottle N/A
N/A
11994911
View Documents
Thermo Scientific Acros
176565000
500 g Glass bottle N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

3240-34-4
322.09
iodobenzene diacetate, diacetoxyiodo benzene, iodosobenzene diacetate, phenyliodine diacetate, phenyliodo diacetate, phenyliodoso acetate, phenyliodosodiacetate, phenyliodoso diacetate, bis acetato phenyliodine, benzene, diacetoxyiodo
[acetyloxy(phenyl)-$l^{3}-iodanyl] acetate
C10H11IO4
ZBIKORITPGTTGI-UHFFFAOYSA-N
76724
CC(=O)OI(C1=CC=CC=C1)OC(=O)C

Specifications

3240-34-4
Colorless to Yellow
Authentic
C10H11IO4
01,508; 03,166; 04,266; 10,214; 13,242
Solubility in water: insoluble
CC(=O)OI(C1=CC=CC=C1)OC(=O)C
322.09
322.09
Solution
161.0°C to 165.0°C
1% max. (60°C)
97.5% min. (Iodometry)
C6H5I(O2CCH3)2
iodobenzene diacetate, diacetoxyiodo benzene, iodosobenzene diacetate, phenyliodine diacetate, phenyliodo diacetate, phenyliodoso acetate, phenyliodosodiacetate, phenyliodoso diacetate, bis acetato phenyliodine, benzene, diacetoxyiodo
ZBIKORITPGTTGI-UHFFFAOYSA-N
[acetyloxy(phenyl)-$l^{3}-iodanyl] acetate
76724
98%
Iodobenzene diacetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 221-808-1

Product Identifier
  • Iodobenzene diacetate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only