missing translation for 'onlineSavingsMsg'
Learn More

Iridium black, 99.8% (metals basis), Thermo Scientific Chemicals

392.00 € - 5804.00 €

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
Synonym ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11336428
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012071.02
0.5 g
392.00 €
0.50g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11346428
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012071.04
2 g
1479.00 €
2g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11356428
View Documents Promotion Details
Thermo Scientific Alfa Aesar
012071.09
10 g
5804.00 €
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The primary use of iridium is as a hardening agent in platinum alloys. Other uses are for the making of crucibles and devices that require high temperatures. Osmium/iridium alloys are used for compass bearings. Iridium is commonly used in complexes like Ir(mppy)3 and other complexes in polymer LED technology to increase the efficiency from 25% to about 100% due to triplet harvesting. Used in high-dose radiation therapy for the treatment of prostate and other forms of cancer. Iridium is used in tips of ballpoint pens. Iridium is used as a catalyst for carbonylation of methanol to create acetic acid. At one time, iridium as an alloy with platinum, was used in bushing the vents of heavy ordnance and, in a finely powdered condition (iridium black) for painting porcelain black.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-88-5
192.22
GKOZUEZYRPOHIO-UHFFFAOYSA-N
23924
iridium
Ir
MFCD00011062
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
CHEBI:49666
[Ir]

Specifications

7439-88-5
MFCD00011062
ion, black, metallicum, iridium, elemental, iridium, ion ir4, iridio, hydrido-iridium, atom, powder, sponge
GKOZUEZYRPOHIO-UHFFFAOYSA-N
iridium
23924
99.8%
Iridium black
Ir
UN3089
Soluble in aqua regia.
[Ir]
192.22
CHEBI:49666
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q

H228-H319-H500

missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-095-9

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only