Learn More
Iron(II) phthalocyanine, 96%, Thermo Scientific Chemicals
Description
Iron phthalocyanine is been investigated as organic electroluminescence materials for the applications in organic solar cells, biosensitizers and display devices such as OLED(Organic Light Emiting Diode), OTFT(Organic Thin Film Transistor), Wearable Display, and e-paper. It is also used in the formation of new type of catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 5 g |
Solubility Information | It is insoluble in water. It is soluble in conc. sulfuric acid. |
Formula Weight | 568.38 |
Percent Purity | 96% |
Physical Form | Crystalline |
Chemical Name or Material | Iron(II) phthalocyanine |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M17L001 | Certificate of Analysis | 21/01/2025 | 039262.06,039262.03 |
R30K026 | Certificate of Analysis | 19/06/2024 | 039262.06,039262.03 |
Q26K045 | Certificate of Analysis | 11/06/2024 | 039262.06,039262.03 |
X03I036 | Certificate of Analysis | 13/10/2022 | 039262.06,039262.03 |
X05I043 | Certificate of Analysis | 13/10/2022 | 039262.06,039262.03 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.