missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 1 kg |
Linear Formula | FeSO4 · 7H2O |
Formula Weight | 278.01g/mol |
Percent Purity | 99.5 to 104.5% (manganometric) |
Grade | puriss. |
Packaging | Plastic Bottle |
Vapor Pressure | 14.6mmHg (25°C) |
Density | 1.898g/mL at 25°C |
Chemical Name or Material | Iron(Ii) Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
M3530 | Certificate of Analysis | 05/04/2023 | 12354-1KG,12354-25KG,12354-4X5KG,12354-500G,12354-5KG |
Safety and Handling

Product Identifier
- Iron(II) sulfate heptahydrate
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Iron(II) sulfate-7-hydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction