missing translation for 'onlineSavingsMsg'
Learn More
Iron(III) Citrate Trihydrate, Pure, Fisher Chemical Product Code.: 10060170

Iron(III) Citrate Trihydrate, Pure, Fisher Chemical

Product Code. 10060170
500 g, Plastic powder jar
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10060170

Brand: Fisher Chemical I/1060/53

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 17217-76-4
Color Brown-Red
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula C6H14FeO10
MDL Number 149617
InChI Key HCQHEELQALBYIW-UHFFFAOYSA-N
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.O.O.O.[Fe]
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;iron;trihydrate
Molecular Weight (g/mol) 302.013
PubChem CID 76420138
Packaging Plastic powder jar
Show More Show Less
Product Identifier
  • Iron(III) citrate trihydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(III) Citrate Trihydrate, Pure, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.