missing translation for 'onlineSavingsMsg'
Learn More
Isatin, 98%, Thermo Scientific Chemicals Product Code.: 11404074

Isatin, 98%, Thermo Scientific Chemicals

Product Code. 11404074
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11404074

Brand: Thermo Scientific Alfa Aesar A12468.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isatin is an endogenous monoamine oxidize (MAO) inhibitor involved in stress and anxiety. Additionally, isatin inhibits alkaline phosphatase (ALP), nitric oxide (NO)-stimulated soluble guanylate cyclase, and other enzymes. Isatins undergo a one-pot Wolff-Kishner like reduction with hydrazine hydrate, under surprisingly mild conditions, to give the corresponding oxindoles. It is used as chromatographic spray reagent for amino acid detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isatin is an endogenous monoamine oxidize (MAO) inhibitor involved in stress and anxiety. Additionally, isatin inhibits alkaline phosphatase (ALP), nitric oxide (NO)-stimulated soluble guanylate cyclase, and other enzymes. Isatins undergo a one-pot Wolff-Kishner like reduction with hydrazine hydrate, under surprisingly mild conditions, to give the corresponding oxindoles. It is used as chromatographic spray reagent for amino acid detection.

Solubility
Soluble in water (1.9 g/L at 20°C).

Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-56-5
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.133
MDL Number MFCD00005718
InChI Key JXDYKVIHCLTXOP-UHFFFAOYSA-N
Synonym isatin, indoline-2,3-dione, 2,3-indolinedione, indole-2,3-dione, isatine, 2,3-dioxoindoline, pseudoisatin, tribulin, isotin, isatic acid lactamShow MoreShow Less
PubChem CID 7054
ChEBI CHEBI:27539
IUPAC Name 1H-indole-2,3-dione
SMILES C1=CC=C2C(=C1)C(=O)C(=O)N2

Specifications

Melting Point ∼199°C to 200°C
pH 7
Flash Point 220°C (428°F)
Odor Odorless
Quantity 500 g
Beilstein 383659
Merck Index 14,5104
Solubility Information Soluble in water (1.9g/L at 20°C).
Formula Weight 147.13
Percent Purity 98%
Chemical Name or Material Isatin
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250378 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA12468.22,A12468.36,A12468.0E
Lot Number10249526 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog NumberA12468.22,A12468.36,A12468.0E
Lot Number10240318 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA12468.22,A12468.36,A12468.0E
Lot NumberB07R042 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12468.22,A12468.36,A12468.0E
Lot Number10191288 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12468.22,A12468.36,A12468.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isatin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.