missing translation for 'onlineSavingsMsg'
Learn More

Hopen™ Iso Air Duckbill Cleanroom Masks
GREENER_CHOICE

ISO Air duckbill cleanroom masks provide a large breathing chamber and are made with the Mass Balance approach from recycled materials.

179.00 € - 180.00 €

Specifications

Product Type Cleanroom Face Mask
Color White
Style Pouch
Certifications/Compliance Grade B
Cleanroom Class Class 1 (ISO 3)
View More Specs

Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17754789
GREENER_CHOICE
View Documents
Hopen™
60.754M
Medium
179.00 €
Pack of 300
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
 
17744789
GREENER_CHOICE
View Documents
Hopen™
60.754L
Large
180.00 €
Pack of 300
Estimated Shipment: 15-01-2025
to see stock.
Add to basket
 
Description

Description

  • Low particle release: Helmke drum tested for use in clean rooms
  • High filtration efficiency of particles, bacteria and viruses
  • Adaptable to users and protocols with two sizes available
  • Highly breathable thanks to a large breathing chamber
  • Softex technology limits risk of particulate contamination while reducing risk of skin irritation for the user
  • Adjustable and malleable nose clip prevents glasses from fogging up
  • ISO AIR range is manufactured in an ISO 8 classified clean room
  • Made in France
Specifications

Specifications

Cleanroom Face Mask
Pouch
Class 1 (ISO 3)
60.754
White
Grade B
Industry
Videos
SDS
Documents

Documents

Product Certifications