missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO Safety Universal Sign 'Wear Laboratory Coat'

Safety Sign to signify that a laboratory coat must be worn.

Brand:  Brady™ 311364

 View more versions of this product

Product Code. 17861775

  • 5.70 € / Pack of 2
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Provide visual warning that laboratory coat must be worn in the area
Specifications

Specifications

M/M059/NT/PE-DIA50-2
Polyester
1.97 in. Dia.
50 mm
Facility Identification, Safety Identification
-40°C to +110°C
-40°F to +230°F
5 x 5 cm
Safety Sign
Gloss
1 Sign
Permanent
Surface Mount
0.099 mm
Round
Universal
Wear Laboratory Coat
White on Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.