missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ISO Safety Universal Sign 'Wear Laboratory Coat'

Safety Sign to signify that a laboratory coat must be worn.

Brand:  Brady™ 311368

 View more versions of this product

Product Code. 17891695

  • 7.85 € / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Provide visual warning that laboratory coat must be worn in the area
Specifications

Specifications

Gloss
1 Sign
Permanent
Surface Mount
0.099 mm
Round
Universal
Wear Laboratory Coat
White on Blue
Polyester
12.4 in. Dia.
315 mm
Facility Identification, Safety Identification
-40°C to +110°C
-40°F to +230°F
31.5 x 31.5 cm
Safety Sign
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers