missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tyvek™ IsoClean™ Boot Covers Product Code.: 15614914
Estimated Shipment: 06-05-2025
to see stock.
Add to basket

DuPont™ Tyvek™ IsoClean™ Boot Covers
BIOPHARMA

Product Code. 15614914 Shop All DuPont Products
Large
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy

Product Code. 15614914

Brand: DuPont™ D15466090

Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. Dupont™ Tyvek™ IsoClean™ Boot Covers provide an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and exceptionally low linting

  • Available in sizes S to XL.
  • Clean-processed and gamma-sterilized.
  • Bound internal seams.
  • Tunnelled elastication at shin and ankles.
  • Ankle ties.
  • Gripper™ soles provide enhanced slip-retardance and robustness.
TRUSTED_SUSTAINABILITY

Specifications

Size Large
Material Tyvek™ IsoClean™
Color White
Disposable Single-use
Sterility Sterile
Sole Type Anti-skid
Seam Type Bound
For Use With (Application) Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics, Other Critical Environments
Certifications/Compliance Clean-processed and sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1), Suitable for use in GMP class A/B (ISO Class 5) clean rooms, PPE Category I
Packaging Quantity 100/Cs.
Packaging Type Individually Packaged in Pairs
Product Type Controlled Environments Boot Covers
Show More Show Less
  • Dupont Tyvek - IsoClean design
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected

      Dupont Tyvek IsoClean design

    • Dupont Tyvek - Packaging for CE certified IsoClean garments
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
       
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected

        Dupont Tyvek Packaging for CE certified IsoClean garments

      • DuPont Tyvek - IsoClean
        Video Player is loading.
        Current Time 0:00
        Duration 0:00
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:00
         
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected

          DuPont Tyvek IsoClean

        Brochures

        Posters

        Product Information

        Certificates

        A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

        5 results found
        Lot Number Certificate Type Date Catalog Number
        Lot NumberAHS2447139 Certificate TypeCertificate of Sterility Date17/12/2024 Catalog NumberD15466090
        Lot NumberAHS2446136 Certificate TypeCertificate of Sterility Date17/12/2024 Catalog NumberD15466090
        Lot NumberAHS2445149 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15466090
        Lot NumberAHS2445162 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15466090
        Lot NumberAHS2440134 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD15466090

        The user should make the risk assessment to determine the protection required.

        Product Content Correction

        Your input is important to us. Please complete this form to provide feedback related to the content on this product.

        Product Title
        DuPont™ Tyvek™ IsoClean™ Boot Covers >

        By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

        Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.