missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ IsoClean™ Boot Covers

Clean-processed to maximize cleanliness and pre-sterilized by gamma irradiation. Dupont™ Tyvek™ IsoClean™ Boot Covers provide an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash. Breathable and exceptionally low linting
276.00 €
Specifications
Material | Tyvek™ IsoClean™ |
---|---|
Color | White |
Disposable | Single-use |
Sterility | Sterile |
Sole Type | Anti-skid |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15604914
![]() |
DuPont™
D15466083 |
Medium |
276.00 €
Pack of 100 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
15614914
![]() |
DuPont™
D15466090 |
Large |
276.00 €
Pack of 100 |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||||
15624914
![]() |
DuPont™
D15466105 |
X-Large |
276.00 €
Pack of 100 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
15694904
![]() |
DuPont™
D15466072 |
Small |
276.00 €
Pack of 100 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
Description
- Available in sizes S to XL.
- Clean-processed and gamma-sterilized.
- Bound internal seams.
- Tunnelled elastication at shin and ankles.
- Ankle ties.
- Gripper™ soles provide enhanced slip-retardance and robustness.
Specifications
Tyvek™ IsoClean™ | |
Single-use | |
Anti-skid | |
Biotech, Pharmaceutical, Medical Device Manufacturing, Food Processing, Cosmetics, Other Critical Environments | |
100/Cs. | |
Controlled Environments Boot Covers |
White | |
Sterile | |
Bound | |
Clean-processed and sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1), Suitable for use in GMP class A/B (ISO Class 5) clean rooms, PPE Category I | |
Individually Packaged in Pairs |
The user should make the risk assessment to determine the protection required.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title