missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers Product Code.: 15379714

DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers

Product Code. 15379714 Shop All DuPont Products
Large
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy

Product Code. 15379714

Brand: DuPont™ D14247987

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Clean-processed and gamma-sterilized. DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash.

  • Hooded coverall includes attached slip-retardant shoe covers
  • Clean-processed and gamma-sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1)
  • Serged internal seams
  • Elastication at wrists, ankles and face
  • Thumb loops prevent sleeves from riding up
  • Zipper closure
  • Aseptically folded
  • White.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size Large
Material HDPE
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Front Zipper
Seam Type Serged
Sterility Sterile
Resistance Liquid, Particle, Chemicals
Certifications/Compliance ISO 11137-1
Model D1424
Show More Show Less
  • Dupont Tyvek - Packaging for CE certified IsoClean garments
    Video Player is loading.
    Current Time 0:00
    Duration 1:40
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:40
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Dupont Tyvek Packaging for CE certified IsoClean garments

  • Dupont Tyvek - IsoClean design
    Video Player is loading.
    Current Time 0:00
    Duration 1:35
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 1:35
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    Dupont Tyvek IsoClean design

  • DuPont Tyvek - IsoClean
    Video Player is loading.
    Current Time 0:00
    Duration 2:33
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 2:33
     
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected

    DuPont Tyvek IsoClean

Brochures

Posters

Product Information

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAHS2447148 Certificate TypeCertificate of Sterility Date17/12/2024 Catalog NumberD14247987
Lot NumberAHS2447141 Certificate TypeCertificate of Sterility Date17/12/2024 Catalog NumberD14247987
Lot NumberAHS2445136 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD14247987
Lot NumberAHS2444111 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD14247987
Lot NumberAHS2446140 Certificate TypeCertificate of Sterility Date11/12/2024 Catalog NumberD14247987
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.