Learn More
DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers
Clean-processed and gamma-sterilized. DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash.
456.00 € - 457.00 €
Specifications
Product Type | Cleanroom Coveralls |
---|---|
Material | HDPE |
Color | White |
Disposable | Single-use |
Ankle Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15309694
|
DuPont™
D14247979 |
Medium | N/A | N/A | N/A | |||||
15379714
|
DuPont™
D14247987 |
Large | N/A | N/A | N/A | |||||
15389734
|
DuPont™
D14247990 |
X-Large |
456.00 €
Pack of 25 |
In Stock
Log in to see stock. ![]() |
||||||
15349754
|
DuPont™
D14248003 |
2X-Large | N/A | N/A | N/A | |||||
Description
Clean-processed and gamma-sterilized. DuPont™ Tyvek™ IsoClean™ Hooded Coverall with Slip-retardant Shoe Covers provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash.
- Hooded coverall includes attached slip-retardant shoe covers
- Clean-processed and gamma-sterilized by gamma-irradiation to SAL of 10-6 (ISO 11137-1)
- Serged internal seams
- Elastication at wrists, ankles and face
- Thumb loops prevent sleeves from riding up
- Zipper closure
- Aseptically folded
- White.
Specifications
Cleanroom Coveralls | |
White | |
Elastic | |
Hooded | |
Serged | |
Liquid, Particle, Chemicals | |
D1424 |
HDPE | |
Single-use | |
Elastic | |
Front Zipper | |
Sterile | |
ISO 11137-1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.