missing translation for 'onlineSavingsMsg'
Learn More

Isohexane, contains <5% N-Hexane, for HPLC

67.70 € - 196.00 €

Chemical Identifiers

CAS 73513-42-5
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD00009406
InChI Key AFABGHUZZDYHJO-UHFFFAOYSA-N
PubChem CID 7892
IUPAC Name 2-methylpentane
SMILES CCCC(C)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10479170
View Documents Promotion Details
Fisher Chemical
H/0405/PB15
1 L Plastic-coated Glass Bottle
67.70 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10214150
View Documents Promotion Details
Fisher Chemical
H/0405/PB17
2.5 L Plastic-coated Glass Bottle
116.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10306380
View Documents Promotion Details
Fisher Chemical
H/0405/21
5 L Aluminum Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

73513-42-5
86.18
AFABGHUZZDYHJO-UHFFFAOYSA-N
2-methylpentane
C6H14
MFCD00009406
7892
CCCC(C)C

Specifications

-154°C
Colorless
Liquid
MFCD00009406
AFABGHUZZDYHJO-UHFFFAOYSA-N
2-methylpentane
7892
73513-42-5
60°C
C6H14
isohexane, pentane, 2-methyl, 2-methylpentan, 2-methyl pentane, dimethylpropylmethane, 1,1-dimethylbutane, iso-hexane, methyl pentane, unii-49ib0u6mld, 2-methyl-pentane
CCCC(C)C
86.18
86.18g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Isohexane

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

SDS
Documents

Documents

Product Certifications