Learn More
Isosorbide, MedChemExpress
MedChemExpress Isosorbide (D-Isosorbide), an orally active vasodilating agent that can be used for the research of heart failure and angina (chest pain). Isosorbide is also an oral hyperosmotic diuretic.
Brand: Medchem Express HY-B1469

Description
Isosorbide (D-Isosorbide), an orally active vasodilating agent that can be used for the research of heart failure and angina (chest pain). Isosorbide is also an oral hyperosmotic diuretic.Specifications
Isosorbide | |
White | |
C6H10O4 | |
D-Isosorbide Dianhydro-D-glucitol | |
O[C@H]1[C@]2([H])[C@]([C@@H](O)CO2)([H])OC1 | |
146.14 | |
Research | |
COVID-19-immunoregulation |
652-67-5 | |
Research | |
5 g | |
H2O : ≥ 100 mg/mL (684.28 mM) ∣DMSO : ≥ 100 mg/mL (684.28 mM) | |
146.14 | |
98.0% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only