missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Isotemp™ CO2 Incubator, 150 L, Stainless Steel

Brand:  Fisherbrand™ 50153430

 View more versions of this product

Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Product Code. 15660667

  • 9330.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Efficient high performance heaters surround the chamber for even temperature distribution
  • Gentle fan assisted air flow pattern is designed for uniform distribution of temperature, CO2, and humidity
  • The 90°C high temperature decontamination cycle simplifies cleaning because no autoclave and toxic chemicals required
  • The cycle eliminates variability in disinfection and has been proven effective against wide range of contaminants including bacteria, molds, and fungal spores and mycoplasma, eliminating the variability in disinfection
  • CO2 sensors are positioned directly in the chamber for precise control. A choice of CO2 sensor technology includes.  
    • Thermal Conductivity (TC) for precise monitoring and reliable long life
    • Infrared (IR) for independent operation for precise, remote monitoring applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RS232
Stainless Steel
TC Sensor
Ambient +0.3° to 55°C
3L panless reservoir
53 x 47 x 60.7 cm
47 cm
60.7 cm
70 kg
230 V
150 L
1 to 20%
<90% at 37°C
CO2 Incubator
Standard
63.7 cm
86.7 cm
53 cm
230 V, 50/60 Hz
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.