Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum is a gas collecting tube with O-ring valves on each end and a centrally located septum.
Brand:  GPE Scientific™ GSS/500
 View more versions of this product
Product Code. 15824774
  • 186.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Cylindrical, oblong shape
  • Gas tight container
  • Gauged volume
  • Standard collecting tube with valved ends
  • Includes centralized septum
Specifications
Specifications
Gas Sample Tube
500 mL