Learn More
GPE Scientific™ J Young Spring Clips for Spherical Joints
GPE Scientific™ J Young Spring Clips for Spherical Joints are spring-based clips designed to securely fasten J Young spherical joints.
4.79 € - 75.20 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
15894774
|
GPE Scientific™
JC13 |
12/2 and 12/5 spherical joints | N/A | N/A | N/A | |||||
15804784
|
GPE Scientific™
JC19 |
18/7 and 18/9 spherical joints |
26.00 €
Each |
Estimated Shipment: 06-06-2025
Log in to see stock. |
||||||
15814784
|
GPE Scientific™
JC29 |
28/12 and 28/15 spherical joints |
4.79 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
15824784
|
GPE Scientific™
JC35 |
35/20 spherical joints | N/A | N/A | N/A | |||||
15834784
|
GPE Scientific™
JC41 |
40/25 spherical joints | N/A | N/A | N/A | |||||
15844784
|
GPE Scientific™
JC51 |
51/35 spherical joints | N/A | N/A | N/A | |||||
Description
- Spring clip design
- Securely fasten J Young spherical joints
Specifications
Spring Clips for J Young Spherical Joints |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
GPE Scientific™ J Young Spring Clips for Spherical Joints