Learn More
GPE Scientific™ J Young Spring Clips for Spherical Joints
GPE Scientific™ J Young Spring Clips for Spherical Joints are spring-based clips designed to securely fasten J Young spherical joints.
Brand: GPE Scientific™ JC19
Description
- Spring clip design
- Securely fasten J Young spherical joints
Specifications
Spring Clips for J Young Spherical Joints | |
18/7 and 18/9 spherical joints |