Learn More
Tristel JET with Spray Head, 2 x 800 mL
JET is a powerful sporicidal foam for the cleaning and disinfection of environmental surfaces and non-porous materials in laboratories.
Brand:  Tristel CCH010201
Product Code. 16658837
  • 94.00 € / Pack of 2
Explore more special offers
Description
Description
  • Compatible with hard, non-porous materials such as laminate, glass, plastic and stainless steel
  • Effective against a wide range of microorganisms within a 1-minute contact time
  • Destroys DNA and RNA in a matter of seconds
  • Complete compliance with EN 14885:2018 and the latest efficacy regulatory requirements
  • Each bottle of JET produces 570 doses of sporicidal foam
  • Can be used with your choice of spreader, which does not have to be plastic
Specifications
Specifications
Foaming Disinfectant
Spray Head
2 Pk.
800 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tristel JET with Spray Head, 2 x 800 mL > 2 x 800 mL