Learn More
Emma Safety Footwear Jordan S1P Denim Shoe
The Jordan is a street inspired safety boot. Its unique heel design is highly supportive and ideal for climbing pallets, ladders and in and out of truck cabs - even when wet.
Brand: Emma Safety Footwear MM95054939
This item is not returnable.
View return policy
Description
- S1P-Safety shoe with sole plate for dry work
- Metal Free
- The Jordan is a street inspired safety boot. Its unique heel design is highly supportive and ideal for climbing pallets, ladders and in and out of truck cabs - even when wet.
- The protective insole protects from sharp objects like nails and shards of metal.
- This sneaker with fiberglass toe cap, is finished with robust denim. DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
- Industries:
- Logistics & transport
- Electronics
- Manufacturing
Specifications
ISO 20345:2011 | |
Blue | |
Denim | |
Denim, Nubuck | |
Standard Width | |
Safety Shoe |
Laces | |
Logistics and Transport, Electronics, Manufacturing | |
Hydro-Tec Antibacterial | |
39 | |
High |
CE_17582596.pdf
File size: 325.6 kB
Product Content Correction
Product Title
Emma Safety Footwear Jordan S1P Denim Shoe > Safety Shoe; Size: 39
Spot an opportunity for improvement?Share a Content Correction