missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Jordan S1P Denim Shoe

The Jordan is a street inspired safety boot. Its unique heel design is highly supportive and ideal for climbing pallets, ladders and in and out of truck cabs - even when wet.

118.00 €

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Blue
For Use With (Application) Logistics and Transport, Electronics, Manufacturing
Lace Material Denim
View More Specs

Products 10
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17582596
View Documents
Emma Safety Footwear
MM95054939
39
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
17592596
View Documents
Emma Safety Footwear
MM95054940
40
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
17502606
View Documents
Emma Safety Footwear
MM95054941
41
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
17512606
View Documents
Emma Safety Footwear
MM95054942
42
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
17522606
View Documents
Emma Safety Footwear
MM95054943
43
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
17532606
View Documents
Emma Safety Footwear
MM95054944
44
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
17542606
View Documents
Emma Safety Footwear
MM95054945
45
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
17552606
View Documents
Emma Safety Footwear
MM95054946
46
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
17562606
View Documents
Emma Safety Footwear
MM95054947
47
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
17572606
View Documents
Emma Safety Footwear
MM95054948
48
118.00 €
1 pair
Estimated Shipment: 17-04-2025
to see stock.
Add to basket
 
Description

Description

  • S1P-Safety shoe with sole plate for dry work
  • Metal Free
  • The Jordan is a street inspired safety boot. Its unique heel design is highly supportive and ideal for climbing pallets, ladders and in and out of truck cabs - even when wet.
  • The protective insole protects from sharp objects like nails and shards of metal.
  • This sneaker with fiberglass toe cap, is finished with robust denim. DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Manufacturing
Specifications

Specifications

ISO 20345:2011
Blue
Denim
Denim, Nubuck
High
Laces
Logistics and Transport, Electronics, Manufacturing
Hydro-Tec Antibacterial
Standard Width
Safety Shoe
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Emma Safety Footwear Jordan S1P Denim Shoe

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.