Learn More
Trajan™ Jumbo Syringe
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
Brand:  Trajan™ 009930
 View more versions of this product
Product Code. 10045470
  • 1405.00 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
  • Designed for holding and dispensing large volumes of gas
  • Heavy duty acrylic barrels
  • Easy access to sample for the addition of standards or removal of subsample via secondary port
  • Plunger stem can be unscrewed for ease of transportation and storage
  • Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifications
Specifications
Acrylic, Acrylic (Barrel)
For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling
Jumbo Syringe
2000MAR-LL-GT
Trajan
130 mm barrel
245 mm
200 mL major scale, 100 mL minor scale
2000 mL
Yes