Learn More
Trajan™ Jumbo Syringe
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
841.00 € - 1405.00 €
Specifications
Material Acrylic, Acrylic (Barrel)
For Use With (Application) For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling
Product Type Jumbo Syringe
View More Specs
Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10751042
View Documents
Trajan™
009920
1000 mL
856.00 €
Each
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
10300811
View Documents
Trajan™
009910
500 mL
841.00 €
Each
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
10045470
View Documents
Trajan™
009930
2000 mL
1405.00 €
Each
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
Description
Description
Hold and dispense large gas volumes. Lightweight and ideal for medical equipment calibration.
  • Designed for holding and dispensing large volumes of gas
  • Heavy duty acrylic barrels
  • Easy access to sample for the addition of standards or removal of subsample via secondary port
  • Plunger stem can be unscrewed for ease of transportation and storage
  • Ideal for calibration of medical equipment such as respirators and spirometers, and for stack and air sampling.
Specifications
Specifications
Acrylic, Acrylic (Barrel)
For calibration of medical equipment such as respirators and spirometers, and for stack and air sampling
Jumbo Syringe