missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Nonmetallic Self-Close Corrosive Safety Containers

Designed to resist acids and caustics

Brand:  Justrite™ 12161

 View more versions of this product

Product Code. 10451165

  • 100.00 € / Each
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Designed to resist acids and caustics

  • Safely holds nonflammable corrosives
  • Translucent white HDPE construction resists dents, punctures, and chemicals
  • Marked in liters, quarts, and gallons for accurate filling and dispensing
  • Flame arrester-free spout allows quick pouring; spring-close cap provides a tight seal
  • Wide mouth on Cat. Nos. 17-985-89B, -89C, and -89D makes filling and pouring easier

†Oval-shaped.

Specifications

Specifications

Stainless Steel Safety Can
1 gal.
White
Spring Close
7.6 in.
4.6 in.
12.75 in.
1.45 in.
Dent, Puncture, Chip Resistant
4 L
Polyethylene
Oval
19.3 cm
11.6 cm
32.3 cm
3.6 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Justrite™ Nonmetallic Self-Close Corrosive Safety Containers > Fittings: Stainless steel; Capacity: 1 gal.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.