Learn More
Justrite™ Nonmetallic Self-Close Corrosive Safety Containers
Designed to resist acids and caustics
100.00 € - 262.00 €
Specifications
Product Type Stainless Steel Safety Can
Color White
Material Polyethylene
Includes Spring Close
Shape Oval
View More Specs
Products 4
Product Code Brand Product Type Capacity (English) Description Price Quantity & Availability  
Product Code Brand Product Type Capacity (English) Description Price Quantity & Availability  
11983669
View Documents
Justrite™
12260
Stainless Steel Safety Can 2.5 gal. Resistant to Dents, Punctures and Chemicals, and Acids This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10451165
View Documents
Justrite™
12161
Stainless Steel Safety Can 1 gal. Dent, Puncture, Chip Resistant
100.00 €
Each
Estimated Shipment: 22-04-2025
to see stock.
Add to basket
11903679
View Documents
Justrite™
12762
Stainless Steel Safety Can 2 gal. Dents, Punctures, Chemicals Resistant This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11913679
View Documents
Justrite™
12765
Stainless Steel Safety Can 5 gal. Dents, Punctures, Chemicals Resistant
262.00 €
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
Description
Description
Designed to resist acids and caustics
  • Safely holds nonflammable corrosives
  • Translucent white HDPE construction resists dents, punctures, and chemicals
  • Marked in liters, quarts, and gallons for accurate filling and dispensing
  • Flame arrester-free spout allows quick pouring; spring-close cap provides a tight seal
  • Wide mouth on Cat. Nos. 17-985-89B, -89C, and -89D makes filling and pouring easier
†Oval-shaped.
Specifications
Specifications
Stainless Steel Safety Can
Polyethylene
Oval
White
Spring Close
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Justrite™ Nonmetallic Self-Close Corrosive Safety Containers