missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Combi Safety Storage Cabinets K-PHOENIX Vol.2-90
Safe and approved storage of aggressive and flammable hazardous substances in working areas
Brand: asecos™ 3033400133838
3517.07 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Alert:
For indoor use only
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one: 2 compartments (vertical seperated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
- User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed (folding door compartment): serial automatic door closing (TSA) releases the arrested door after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible) / cylinder lock
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- GS certified
- CE compliant
Services
Five years
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
617 mm | |
Left: 524 x 450 x 1742 mm, Top Right: 542 x 491 x 862 mm, Bottom Right: 542 x 491 x 862 mm | |
Wing, Folding, Auto-close | |
1969 mm | |
K90.196.120.MF.FWAC | |
7 | |
Powder-coated plain | |
1197 mm | |
Light Gray | |
Combi safety storage cabinets |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), 4 x pull-out shelf | |
524 mm | |
617 x 1197 x 1969 mm | |
Safe and approved storage of aggressive and flammable hazardous substances in working areas | |
1742 mm | |
2 | |
Pull-out | |
541 kg | |
450 mm | |
Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction