missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ K70 Silicon Hoses

Kartell™ K70 Silicon Hoses are constructed of silicon, making them a transparent and flexible connection to various components and accessories.

Brand:  Kartell™ 0392200

 View more versions of this product

Product Code. 15712855

  • 20.00 € / 10m
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Kartell™ K70 Silicon Hoses are constructed of silicon, making them a transparent and flexible connection to various components and accessories.

  • Flexible and transparent
  • FDA, USP, Class VI, A 50p60, EPC VI.1.3.2 approved
  • Can withstand temperatures of -60°C to 200°C
Specifications

Specifications

FDA, USP, Class VI, A 50p60 and EPC VI.1.3.2 Approved
Silicone
10 m
Transparent
3 mm
5 mm
-60°C
200°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.