missing translation for 'onlineSavingsMsg'
Learn More

Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical™

Single-component reagent; Pyridine free

212.00 € - 348.00 €

Specifications

Purity or Free of Statement Pyridine free
Chemical Name or Material Aqualine™ Complete 5
Color Brown
Name Note Karl Fischer Reagent
CAS 111-90-0, 288-32-4, 7446-09-5, 7553-09-5
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10181570
View Documents
Fisher Chemical
K/2000/15
1 L
212.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10676262
View Documents
Fisher Chemical
K/2000/17
2.5 L
348.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Single-component reagent; Pyridine free

  • Single-component volumetric reagent for high water content analysis
  • Water equivalent: 5.0mg H2O/mL
  • Unlimited water capacity
  • Perform titer determination before use
  • Pyridine free
Specifications

Specifications

Pyridine free
Brown
111-90-0, 288-32-4, 7446-09-5, 7553-09-5
Liquid
Aqualine™ Complete 5
Karl Fischer Reagent
5.2mg of water/mL min.
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.