missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical™
Single-component reagent; Pyridine free
Brand: Fisher Chemical K/2000/17
254.94 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
Single-component reagent; Pyridine free
- Single-component volumetric reagent for high water content analysis
- Water equivalent: 5.0mg H2O/mL
- Unlimited water capacity
- Perform titer determination before use
- Pyridine free

Specifications
Aqualine™ Complete 5 | |
Karl Fischer Reagent | |
2.5 L | |
5.2mg of water/mL min. | |
Liquid |
Brown | |
Pyridine free | |
111-90-0, 288-32-4, 7446-09-5, 7553-09-5 | |
Amber glass bottle | |
81734 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Aqualine Complete 5
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H360D-May damage the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Karl Fischer Aqualine™ Complete 5, for Karl Fischer Titration By Volumetry, Fisher Chemical™ > 2.5L, Amber glass bottle
Spot an opportunity for improvement?Share a Content Correction