Learn More
kb-NB77-78, MedChemExpress
MedChemExpress kb-NB77-78 is an analogue of CID797718, but shows no PKD inhibitory activity.

167.40 € - 267.30 €
Specifications
Chemical Name or Material | kb-NB77-78 |
---|---|
CAS | 1350622-33-1 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C18H25NO3Si |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16477138
![]() |
Medchem Express
HY-16698 |
10 mg |
267.30 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16419677
![]() |
Medchem Express
HY-16698 |
5 mg |
167.40 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
kb-NB77-78 is an analogue of CID797718, but shows no PKD inhibitory activity.Specifications
kb-NB77-78 | |
Light Yellow | |
C18H25NO3Si | |
O=C1OC2=CC=C(O[Si](C)(C(C)(C)C)C)C=C2C3=C1NCCC3 | |
331.48 | |
Research | |
Cancer-programmed cell death |
1350622-33-1 | |
Research | |
DMSO : 25 mg/mL (75.42 mM; Need ultrasonic) | |
331.48 | |
99.97% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.