Learn More
kb-NB77-78, MedChemExpress
MedChemExpress kb-NB77-78 is an analogue of CID797718, but shows no PKD inhibitory activity.
Brand: Medchem Express HY-16698
This item is not returnable.
View return policy
Description
kb-NB77-78 is an analogue of CID797718, but shows no PKD inhibitory activity.Specifications
kb-NB77-78 | |
Light Yellow | |
C18H25NO3Si | |
DMSO : 25 mg/mL (75.42 mM; Need ultrasonic) | |
331.48 | |
99.97% | |
Solid |
1350622-33-1 | |
Research | |
5 mg | |
O=C1OC2=CC=C(O[Si](C)(C(C)(C)C)C)C=C2C3=C1NCCC3 | |
331.48 | |
Research | |
Cancer-programmed cell death |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
kb-NB77-78, MedChemExpress