missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ KCl 3M Electrode Filling Solution

Equipped with a special cap to facilitate easy filling of electrodes. Mettler Toledo™ KCl 3M Electrode Filling Solution is an electrolyte solution that enables optimum performance of pH and ion-selective electrodes. This bridge electrolyte contains no silver ions than can contaminate samples.

Brand:  Mettler Toledo™

25.99 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.


 View more versions of this product

Product Code. 10780412

  • 31.30 € / 250mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

An electrode is the main actor of pH and ion measurement. They require special care and handling. The electrode should remain filled with fresh, clean electrolyte solution. Regularly refilling or replacing the electrolyte solution is necessary to maintain good electrode performance.

The reference electrode of a pH or Ion-Selective Electrode (ISE) should be filled only with the recommended electrolyte solution. The correct electrolyte minimizes junction potentials and provides optimum temperature and time response.

3mol/L Potassium Chloride solution is appropriate for most applications.

  • Used by most Mettler Toledo pH sensors, due to the ARGENTHAL™ reference system
  • DX-series ISE half-cells can also use a 3M KCl electrolyte bridge solution
  • No risk of contaminating samples with silver ions
  • Equipped with a special cap to facilitate easy filling of electrodes
  • Available in 25mL and 250mL bottles, and a 6 × 250mL bulk pack
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Colorless
ARGENTHAL™ Reference System pH Electrodes, DX Series ISE Half Cells
Yes
Neutral
3 mol/L KCl
Potassium Chloride Electrolyte
Bottle with Special Filling Cap
250 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mettler Toledo™ KCl 3M Electrode Filling Solution > Quantity: 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.