missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ KCl 3M Electrode Filling Solution

Equipped with a special cap to facilitate easy filling of electrodes. Mettler Toledo™ KCl 3M Electrode Filling Solution is an electrolyte solution that enables optimum performance of pH and ion-selective electrodes. This bridge electrolyte contains no silver ions than can contaminate samples.

9.55 € - 110.00 €

Specifications

Color Colorless
Concentration 3 mol/L KCl
For Use With (Equipment) ARGENTHAL™ Reference System pH Electrodes, DX Series ISE Half Cells
NIST Traceable Yes
Packaging Bottle with Special Filling Cap
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11907781
View Documents
Mettler Toledo™
51343180
25 mL
9.55 €
25mL
In Stock
to see stock.
Edge
Add to basket
 
11932535
View Documents
Mettler Toledo™
51350080
6 x 250 mL
110.00 €
1500mL
In Stock
to see stock.
Edge
Add to basket
 
10780412
View Documents
Mettler Toledo™
51350072
250 mL
31.30 €
250mL
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

An electrode is the main actor of pH and ion measurement. They require special care and handling. The electrode should remain filled with fresh, clean electrolyte solution. Regularly refilling or replacing the electrolyte solution is necessary to maintain good electrode performance.

The reference electrode of a pH or Ion-Selective Electrode (ISE) should be filled only with the recommended electrolyte solution. The correct electrolyte minimizes junction potentials and provides optimum temperature and time response.

3mol/L Potassium Chloride solution is appropriate for most applications.

  • Used by most Mettler Toledo pH sensors, due to the ARGENTHAL™ reference system
  • DX-series ISE half-cells can also use a 3M KCl electrolyte bridge solution
  • No risk of contaminating samples with silver ions
  • Equipped with a special cap to facilitate easy filling of electrodes
  • Available in 25mL and 250mL bottles, and a 6 × 250mL bulk pack
Specifications

Specifications

Colorless
ARGENTHAL™ Reference System pH Electrodes, DX Series ISE Half Cells
Bottle with Special Filling Cap
3 mol/L KCl
Yes
Neutral
Videos
SDS
Documents

Documents

Product Certifications