missing translation for 'onlineSavingsMsg'
Learn More

KD Scientific Legato Syringe Pumps: Racks

For use with Legato Syringe Pumps

Brand:  KD Scientific 788300

 View more versions of this product

Product Code. 11985586

  • 975.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Accommodates up to ten 0.5 μL to 20 mL syringes or six 30 to 60 mL plastic or glass syringes

Certifications

CE Marked, UL Listed, CSA, WEE, CB scheme, EU RoHS

Specifications

Specifications

0/1 V
CE, UL, CSA, WEE, CB scheme, EU RoHS
Legato syringe pump
0.5 μL to 140 mL
Large Syringe Multi-rack
±0.35%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.